COM448632677 Combi-Blocks, Inc.

1-[7-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran-2-yl]ethan-1-one

  • MDL number MFCD24039605
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1375
Availability details for
Quantity
  • Availability
  • Add to Cart