COM448632697 Combi-Blocks, Inc.

1-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)phenyl]pyrrolidin-2-one

  • MDL number MFCD24039708
  • Cas Number : 2223041-71-0
  • Molecular Weight : 355.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1395

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1395
Availability details for
Quantity
  • Availability
  • Add to Cart