COM448632701 Combi-Blocks, Inc.

1-[5-Methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]propan-1-one

  • MDL number MFCD27936299
  • Cas Number : 1777825-83-8
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1399
Availability details for
Quantity
  • Availability
  • Add to Cart