COM448632706 Combi-Blocks, Inc.

tert-Butyl 7-(tetramethyl-1,3,2-dioxaborolan-2-yl)indole-1-carboxylate

  • MDL number MFCD27936304
  • Cas Number : 2281870-44-6
  • Molecular Weight : 343.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1404
Availability details for
Quantity
  • Availability
  • Add to Cart