COM448632711 Combi-Blocks, Inc.

4,5-Difluoro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-1,3-benzodiazole

  • MDL number MFCD27936307
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1410
Availability details for
Quantity
  • Availability
  • Add to Cart