COM448632712 Combi-Blocks, Inc.

6,7-Difluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-1,2,3-benzotriazole

  • MDL number MFCD24039391
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1411
Availability details for
Quantity
  • Availability
  • Add to Cart