COM448632716 Combi-Blocks, Inc.

1-Ethyl-7-fluoro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-quinoxaline-2,3-dione

  • MDL number MFCD27936310
  • Cas Number : 2377611-66-8
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1415
Availability details for
Quantity
  • Availability
  • Add to Cart