COM448632718 Combi-Blocks, Inc.

1-Methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)indazol-3-amine

  • MDL number MFCD27936312
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1417
Availability details for
Quantity
  • Availability
  • Add to Cart