COM448632722 Combi-Blocks, Inc.

Ethyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)hexanoate

  • MDL number MFCD27936315
  • Cas Number : 2243982-06-9
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1421
Availability details for
Quantity
  • Availability
  • Add to Cart