COM448632729 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-[(2-nitrophenyl)methyl]-1,3,2-dioxaborolane

  • MDL number MFCD27936321
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1428
Availability details for
Quantity
  • Availability
  • Add to Cart