COM448632733 Combi-Blocks, Inc.

Methyl (2E)-3-{4-[(tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]phenyl}prop-2-enoate

  • MDL number MFCD27936325
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1432
Availability details for
Quantity
  • Availability
  • Add to Cart