COM448632739 Combi-Blocks, Inc.

Methyl 3-methoxy-4-[(tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]benzoate

  • MDL number MFCD27936331
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1438
Availability details for
Quantity
  • Availability
  • Add to Cart