COM448632758 Combi-Blocks, Inc.

2-Propenoic acid, 3-[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-, (2e)-

  • MDL number MFCD22493999
  • Cas Number : 1001200-58-3
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1462
Availability details for
Quantity
  • Availability
  • Add to Cart