COM448632763 Combi-Blocks, Inc.

2-Nitro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD27936346
  • Cas Number : 2227082-58-6
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1467

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1467
Availability details for
Quantity
  • Availability
  • Add to Cart