COM448632767 Combi-Blocks, Inc.

2-[5-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid

  • MDL number MFCD24039845
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1471
Availability details for
Quantity
  • Availability
  • Add to Cart