COM448632771 Combi-Blocks, Inc.

3-Amino-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol

  • MDL number MFCD26516757
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1475

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1475
Availability details for
Quantity
  • Availability
  • Add to Cart