COM448632780 Combi-Blocks, Inc.

5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine-2,3-diamine

  • MDL number MFCD27936353
  • Cas Number : 1350755-71-3
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1485
Availability details for
Quantity
  • Availability
  • Add to Cart