COM448632785 Combi-Blocks, Inc.

2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD23380352
  • Cas Number : 1400976-18-2
  • Molecular Weight : 316.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1490
Availability details for
Quantity
  • Availability
  • Add to Cart