COM448632795 Combi-Blocks, Inc.

5-Methyl-3-nitro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD27936362
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1500
Availability details for
Quantity
  • Availability
  • Add to Cart