COM448632804 Combi-Blocks, Inc.

(2S)-2-[[(tert-Butoxy)carbonyl]amino]-3-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanoic acid

  • MDL number MFCD24039657
  • Cas Number : 2377587-64-7
  • Molecular Weight : 391.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1513
Availability details for
Quantity
  • Availability
  • Add to Cart