COM448632808 Combi-Blocks, Inc.

7-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,4-dihydro-1h-3,1-benzoxazine-2,4-dione

  • MDL number MFCD24039388
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1520
Availability details for
Quantity
  • Availability
  • Add to Cart