COM448632810 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-(2-methyl-6-nitrophenyl)-1,3,2-dioxaborolane

  • MDL number MFCD27936369
  • Cas Number : 2361310-64-5
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1522
Availability details for
Quantity
  • Availability
  • Add to Cart