COM448632814 Combi-Blocks, Inc.

Acetamide, n-[5-chloro-4-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-

  • MDL number MFCD23381017
  • Cas Number : 1374606-62-8
  • Molecular Weight : 309.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1526

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1526
Availability details for
Quantity
  • Availability
  • Add to Cart