COM448632816 Combi-Blocks, Inc.

Benzenesulfonamide, N-(2-aminoethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

  • MDL number MFCD09027067
  • Cas Number : 1228148-98-8
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1528
Availability details for
Quantity
  • Availability
  • Add to Cart