COM448632820 Combi-Blocks, Inc.

2-Methyl-5-sulfamoylphenylboronic acid pinacol ester

  • MDL number MFCD18730620
  • Cas Number : 909187-69-5
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1532
Availability details for
Quantity
  • Availability
  • Add to Cart