COM448632824 Combi-Blocks, Inc.

2,3-Dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline hydrochloride

  • MDL number MFCD27936378
  • Molecular Weight : 283.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1536

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1536
Availability details for
Quantity
  • Availability
  • Add to Cart