COM448632840 Combi-Blocks, Inc.

[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]thiourea

  • MDL number MFCD27936390
  • Cas Number : 2249914-86-9
  • Molecular Weight : 278.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1553
Availability details for
Quantity
  • Availability
  • Add to Cart