COM448632850 Combi-Blocks, Inc.

N-Cyclopropyl-4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]benzenesulfonamide

  • MDL number MFCD27936400
  • Molecular Weight : 389.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1563
Availability details for
Quantity
  • Availability
  • Add to Cart