COM448632855 Combi-Blocks, Inc.

6-Fluoro-3-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

  • MDL number MFCD22414868
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1568
Availability details for
Quantity
  • Availability
  • Add to Cart