COM448632858 Combi-Blocks, Inc.

3-Ethoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD24039826
  • Cas Number : 1801514-64-6
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1571
Availability details for
Quantity
  • Availability
  • Add to Cart