COM448632864 Combi-Blocks, Inc.

3-Chloro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxylic acid

  • MDL number MFCD27936403
  • Molecular Weight : 283.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1577
Availability details for
Quantity
  • Availability
  • Add to Cart