COM448632866 Combi-Blocks, Inc.

Methyl([2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethyl])amine

  • MDL number MFCD18734051
  • Cas Number : 2377610-45-0
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1579
Availability details for
Quantity
  • Availability
  • Add to Cart