COM448632868 Combi-Blocks, Inc.

1-[2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidin-2-one

  • MDL number MFCD24039949
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1581
Availability details for
Quantity
  • Availability
  • Add to Cart