COM448632875 Combi-Blocks, Inc.

N-Cyclohexyl-2-ethoxy-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD24039465
  • Molecular Weight : 373.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1588
Availability details for
Quantity
  • Availability
  • Add to Cart