COM448632877 Combi-Blocks, Inc.

(E)-N'-Hydroxy-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1-carboximidamide

  • MDL number MFCD27936405
  • Cas Number : 2088368-01-6
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1590
Availability details for
Quantity
  • Availability
  • Add to Cart