COM448632879 Combi-Blocks, Inc.

tert-Butyl 4-[2-oxo-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethyl]piperazine-1-carboxylate

  • MDL number MFCD24039358
  • Molecular Weight : 430.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1592
Availability details for
Quantity
  • Availability
  • Add to Cart