COM448632886 Combi-Blocks, Inc.

4-Butyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2h-1,4-benzoxazin-3-one

  • MDL number MFCD24039753
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1599
Availability details for
Quantity
  • Availability
  • Add to Cart