COM448632889 Combi-Blocks, Inc.

1-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbohydrazide

  • MDL number MFCD24039916
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1602
Availability details for
Quantity
  • Availability
  • Add to Cart