COM448632897 Combi-Blocks, Inc.

methyl 1-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carboxylate

  • MDL number MFCD18761349
  • Cas Number : 1004294-83-0
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1610
Availability details for
Quantity
  • Availability
  • Add to Cart