COM448632898 Combi-Blocks, Inc.

2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)propyl)isoindoline-1,3-dione

  • MDL number MFCD22987683
  • Cas Number : 1704068-63-2
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1611
Availability details for
Quantity
  • Availability
  • Add to Cart