COM448632902 Combi-Blocks, Inc.

(2R)-1-Hydroxy-S-(4-methylphenyl)-N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}propane-2-sulfonamido

  • MDL number MFCD27936418
  • Molecular Weight : 445.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1616
Availability details for
Quantity
  • Availability
  • Add to Cart