COM448632903 Combi-Blocks, Inc.

2-[2-(4-Fluoro-2-nitrophenoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936419
  • Molecular Weight : 359.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1617
Availability details for
Quantity
  • Availability
  • Add to Cart