COM448632904 Combi-Blocks, Inc.

4-(2-Fluoro-4-nitrophenoxy)phenylboronic acid pinacol ester

  • MDL number MFCD27936420
  • Cas Number : 2377606-39-6
  • Molecular Weight : 359.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1618
Availability details for
Quantity
  • Availability
  • Add to Cart