COM448632909 Combi-Blocks, Inc.

3-Fluoro-4-[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]aniline hydrochloride

  • MDL number MFCD27936425
  • Molecular Weight : 365.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1623
Availability details for
Quantity
  • Availability
  • Add to Cart