COM448632915 Combi-Blocks, Inc.

1-[4-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine

  • MDL number MFCD18764905
  • Cas Number : 2377607-17-3
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1629
Availability details for
Quantity
  • Availability
  • Add to Cart