COM448632920 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,3]thiazolo[4,5-b]pyridine-2-thiol

  • MDL number MFCD27936433
  • Molecular Weight : 294.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1634
Availability details for
Quantity
  • Availability
  • Add to Cart