COM448632926 Combi-Blocks, Inc.

2-chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide

  • MDL number MFCD18723978
  • Molecular Weight : 282.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1640
Availability details for
Quantity
  • Availability
  • Add to Cart