COM448632928 Combi-Blocks, Inc.

tert-Butyl N-cyclopropyl-N-[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]carbamate

  • MDL number MFCD27936439
  • Molecular Weight : 361.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1642

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1642
Availability details for
Quantity
  • Availability
  • Add to Cart