COM448632932 Combi-Blocks, Inc.

5-Chloro-6-methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD27936443
  • Molecular Weight : 268.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1646
Availability details for
Quantity
  • Availability
  • Add to Cart