COM448632936 Combi-Blocks, Inc.

3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)pyridine-2-thiol

  • MDL number MFCD27936447
  • Molecular Weight : 305.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1650

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1650
Availability details for
Quantity
  • Availability
  • Add to Cart